Learn More
Medchemexpress LLC HY-15461 50mg , Ertugliflozin CAS:1210344-57-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1546150MG
Encompass_Preferred
HY-15461 50mg Ertugliflozin CAS: 1210344-57-2 Ertugliflozin (PF-04971729) is a potent, selective and orally active inhibitor of the sodium-dependent glucose cotransporter 2 (SGLT2), with an IC50 of 0.877 nM for h-SGLT2. Has the potential for the treatment of type 2 diabetes mellitus. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0915