Learn More
Medchemexpress LLC Ertugliflozin L-pyroglutamic acid | 1210344-83-4 | 99.8% | 566.00 | 5 MG
SDP
Catalog No. 5000364180
Encompass_Preferred
Ertugliflozin L-pyroglutamic acid is a potent, selective, and orally active inhibitor of the sodium-dependent glucose cotransporter 2 (SGLT2), with an IC50 of 0.877 nM for h-SGLT2. It has potential for the treatment of type 2 diabetes mellitus.
  • Potent, selective, and orally active inhibitor of the sodium-dependent glucose cotransporter 2 (SGLT2)
  • Potential for the treatment of type 2 diabetes mellitus
  • Demonstrates >2000-fold selectivity for SGLT2 inhibition (relative to SGLT1) in vitro
  • Reveals a concentration-dependent glucosuria after oral administration to rats
Catalog No. 50-003-64180 Supplier Medchemexpress LLC Supplier No. 5MGHY15461A
Ertugliflozin L-pyroglutamic acid is a potent, selective, and orally active inhibitor of the sodium-dependent glucose cotransporter 2 (SGLT2), with an IC50 of 0.877 nM for h-SGLT2. It has potential for the treatment of type 2 diabetes mellitus.
  • Potent, selective, and orally active inhibitor of the sodium-dependent glucose cotransporter 2 (SGLT2)
  • Potential for the treatment of type 2 diabetes mellitus
  • Demonstrates >2000-fold selectivity for SGLT2 inhibition (relative to SGLT1) in vitro
  • Reveals a concentration-dependent glucosuria after oral administration to rats