Learn More
Medchemexpress LLC HY-N7109 100mg Medchemexpress, Erucic acid CAS:112-86-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN7109100MG
Encompass_Preferred
Medchemexpress, HY-N7109 100mg Erucic acid CAS:112-86-7 Erucic acid, a monounsaturated fatty acid (MUFA), is isolated from the seed of Raphanus sativus L. Erucic acid can readily cross the blood-brain barrier (BBB), it has been reported to normalize the accumulation of very long-chain fatty acids in the brain. Erucic acid can improve cognitive impairment and be effective against dementia [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3983