Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erucic acid, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012514 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2514 1 g
11-101-2515 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-2514 Supplier MilliporeSigma Supelco Supplier No. 456291GF
Add to Cart
Add to Cart

Specifications

CAS 29232-93-7
Flash Point 113°C
Physical Form Neat
Linear Formula CH3(CH2)7 CH=CH(CH2)11 COOH
MDL Number MFCD00063188
Melting Point 28°C to 32°C
Boiling Point 358°C (400 mmHg)
Synonym cis-13-Docosenoic acid; Prifac 2990
Type Analytical Standard
Quantity 1 g
Formula Weight 338.57
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.