Learn More
Biomatik Corporation Ervfrd-1 Polyclonal Antibody
Supplier:  Biomatik Corporation CAC10866
Encompass_Preferred
Ervfrd-1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to ERVFRD-1. The Ervfrd-1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Syncytin-2 Protein (16-250Aa). Applications: ELISA, WB, IHC. Target Synonyms: ERVFRD-1. Accession Number: P60508. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7680