Learn More
Innovating Science™ Erycard™ ABO/Rh Blood Typing Card
SCI_ED
Catalog No. S001210 Shop All Aldon Products
Change view
Click to view available options
Test Time:
5 Minutes
Catalog No. Test Time
S001210 5 Minutes
1 options
Catalog No. S001210 Supplier Innovating Science™ Supplier No. IS3181
Add to Cart
Add to Cart
Use the Erycard™ Blood Type Test Kit to determine ABO/Rh blood type of real human blood in under five minutes.
  • Determine ABO/Rh blood type of real human blood in under five minutes.
  • A small amount of blood is applied to a series of wells containing lyophilized blood typing antibodies, followed by the addition of a wash buffer that flows across each well.
  • Red blood cells that agglutinate in the presence of a particular antibody will not be rinsed from the sample well and the well will remain red, indicating a positive result.
  • Red blood cells that have not undergone agglutination will be rinsed from the well and the well will revert to white, indicating a negative result.
  • Each card contains a negative control well to ensure accurate results.
  • Includes 24 trays, wash buffer, and blood droppers.
Specifications
CE Marker No
Detectable Analytes ABO Antigen, Rh D antigen
Type Card
Includes 24 Trays, Wash Buffer, and Blood Droppers
Test Time 5 Minutes
Form Ready-to-Use (RTU)