Learn More
Medchemexpress LLC HY-N7079 500mg Medchemexpress, Erythorbic acid CAS:89-65-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN7079500MG
Encompass_Preferred
Medchemexpress, HY-N7079 500mg Erythorbic acid CAS:89-65-6 Erythorbic acid (D-Isoascorbic acid), produced from sugars derived from different sources, such as beets, sugar cane, and corn, is a food additive used predominantly in meats, poultry, and soft drinks. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2982