Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Erythorbic acid (standard) | 89-65-6 | 50 MG
SDP

Catalog No. 5000418858
Encompass_Preferred

Erythorbic acid (Standard) is the analytical standard of Erythorbic acid (HY-N7079). It is intended for research and analytical applications. Erythorbic acid is an orally active compound, improves the radiation resistance of *Listeria monocytogenes*, reacts with free radicals to reduce oxidative damage, and has weak antiscorbic activity. It can also prevent oxidative discoloration in meat products.

  • Purity: 99.96%
  • Application: Analytical standard for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Molecular weight: 176.12
  • Formula: C6H8O6
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Initial source: Microorganisms
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: Store under recommended conditions in the certificate of analysis.

Catalog No. 50-004-18858 Supplier Medchemexpress LLC Supplier No. HYN7079R50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Erythorbic acid (Standard) is the analytical standard of Erythorbic acid (HY-N7079). It is intended for research and analytical applications. Erythorbic acid is an orally active compound, improves the radiation resistance of *Listeria monocytogenes*, reacts with free radicals to reduce oxidative damage, and has weak antiscorbic activity. It can also prevent oxidative discoloration in meat products.

  • Purity: 99.96%
  • Application: Analytical standard for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Molecular weight: 176.12
  • Formula: C6H8O6
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Initial source: Microorganisms
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: Store under recommended conditions in the certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.