Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Erythorbic acid (Standard) | 89-65-6 | 500 MG
SDP

Catalog No. 5000418678
Encompass_Preferred

Erythorbic acid (Standard) is an analytical standard intended for research and analytical applications. It is an orally active compound that improves the radiation resistance of *Listeria monocytogenes* and can react with free radicals to reduce oxidative damage. It has weak antiscorbic activity and helps prevent oxidative discoloration in meat products.

  • Purity of 99.96%
  • Molecular weight of 176.12
  • Formula C6H8O6
  • Solid appearance, light yellow to yellow color
  • Derived from microorganisms
  • Suitable for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS

Catalog No. 50-004-18678 Supplier Medchemexpress LLC Supplier No. HYN7079R500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Erythorbic acid (Standard) is an analytical standard intended for research and analytical applications. It is an orally active compound that improves the radiation resistance of *Listeria monocytogenes* and can react with free radicals to reduce oxidative damage. It has weak antiscorbic activity and helps prevent oxidative discoloration in meat products.

  • Purity of 99.96%
  • Molecular weight of 176.12
  • Formula C6H8O6
  • Solid appearance, light yellow to yellow color
  • Derived from microorganisms
  • Suitable for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.