Learn More
Bioworld Erythrina cristagalli (Coral tree) Lectin (ECL/ECA) - FITC
Supplier:  Bioworld 217610182
Encompass
Technical Specifications:Ligand Source:Erythrina cristagalliMolecular Weight: 54 kDaCarbohydrate Specificity:Galβ1,4GlcNAcInhibiting/Eluting Sugar:LactoseCarbohydrate for Elution: 0.2MLactoseBlood Group Specificity: A (-SA)Conjugate: FITCForm: LiquidIsoelectric (pI): 6.3-6.5Storage: -20°CApplication note: Calcium, Manganese, and Zinc are required for binding.
Catalog No. 50-199-122