Learn More
Bioworld Erythrina cristagalli (Coral tree) Lectin (ECL/ECA) - Rhodamine (TRITC)
Supplier:  Bioworld 217610191
Encompass
Technical Specifications:Ligand Source:Erythrina cristagalliMolecular Weight:54 kDaCarbohydrate Specificity:Galβ1,4GlcNAcInhibiting/Eluting Sugar:LactoseCarbohydrate for Elution:0.2MLactoseBlood Group Specificity:A (-SA)Conjugate: RhodamineForm:LiquidIsoelectric (pI):6.3-6.5Storage: -20°CApplication note: Calcium, Manganese, and Zinc are required for binding.
Catalog No. 50-199-123