Learn More
Bioworld ERYTHRINA CRISTAGALLICORA
Supplier:  Bioworld 201200061
Encompass
ERYTHRINA CRISTAGALLI (CORAL TREE) LECTIN SEPAROPORE 4B-CL: Erythrina cristagalli (coral tree) lectin is a 54,000 dalton glycoprotein consisting of two different subunits of approximately 28,000 and 26,000 daltons. It has a specificity t...
Catalog No. 50-197-521