Learn More
Apexbio Technology LLC Erythritol 149-32-6 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B17475.1
Encompass_Preferred
Erythritol (CAS 149-32-6) is a four-carbon polyol commonly studied for applications as a low-calorie sweetener Structurally related to other sugar alcohols it exhibits minimal caloric contribution due to limited systemic metabolism and absorption Mechanistically erythritol provides sweetness by interacting with taste receptors on the tongue but it does not significantly elevate blood glucose or insulin levels making it a relevant compound for diabetes and metabolic research Emerging evidence also suggests potential effects on gut microbiota and vascular health warranting exploration in biomedical studies focused on metabolism and cardiovascular function
Catalog No. 50-000-61989