Learn More
Medchemexpress LLC Erythro-guaiacylglycerol-β-ferulic acid ether | 1372162-61-2 | 390.38 | 1 MG
SDP
Catalog No. 5000415065
Encompass_Preferred
Erythro-guaiacylglycerol-β-ferulic acid ether is a natural product derived from *Gueldenstaedtia verna*. It is intended for research applications.
  • Isolated from *Gueldenstaedtia verna*
  • Suitable for research use
  • Store at 4°C, protect from light
  • In solvent: -80°C for 6 months; -20°C for 1 month, protect from light
Catalog No. 50-004-15065 Supplier Medchemexpress LLC Supplier No. HYN115161MG
Erythro-guaiacylglycerol-β-ferulic acid ether is a natural product derived from *Gueldenstaedtia verna*. It is intended for research applications.
  • Isolated from *Gueldenstaedtia verna*
  • Suitable for research use
  • Store at 4°C, protect from light
  • In solvent: -80°C for 6 months; -20°C for 1 month, protect from light