Learn More
Cayman Chemical Erythrodiol, Purity: ≥95%, CAS: 545-48-2, 50 mg
SDP
Catalog No. 502978758
Encompass_Preferred
A triterpene with diverse biological activities; induces relaxation of isolated rat aortic rings precontracted with phenylephrine (EC50 = 3.38 µM); inhibits the growth of HT-29 adenocarcinoma cells (EC50 = 48.8 µM); induces production of ROS and apoptosis in MCF-7 cells at 100 µM; topical administration reduces TPA-induced ear edema and MPO activity in mice at 0.5 mg/ear
Catalog No. 50-297-8758 Supplier Cayman Chemical Supplier No. 3013550 MG
A triterpene with diverse biological activities; induces relaxation of isolated rat aortic rings precontracted with phenylephrine (EC50 = 3.38 µM); inhibits the growth of HT-29 adenocarcinoma cells (EC50 = 48.8 µM); induces production of ROS and apoptosis in MCF-7 cells at 100 µM; topical administration reduces TPA-induced ear edema and MPO activity in mice at 0.5 mg/ear