Learn More
TARGETMOL CHEMICALS INC ERYTHRODIOL 5MG
SDP
Catalog No. 502576259
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Erythrodiol an olive oil component promotes Cholesterol efflux (ChE) by selectively inhibiting the degradation of ABCA1 protein. purity: 99%
Catalog No. 50-257-6259 Supplier TARGETMOL CHEMICALS INC Supplier No. T81655MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Erythrodiol an olive oil component promotes Cholesterol efflux (ChE) by selectively inhibiting the degradation of ABCA1 protein. purity: 99%