Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erythrodiol, ≥97% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014563 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4563 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4563 Supplier MilliporeSigma Supelco Supplier No. 0925810MGF
Add to Cart
Add to Cart

Erythrodiol is a natural triterpenoid produced in olives. It is believed to exhibit antiproliferative and proapoptotic activity in colon adenocarcinoma cells.

Featured Industries: Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

CAS 545-48-2
Linear Formula C30H50O2
MDL Number MFCD00017294
Melting Point 230°C to 231°C
Synonym Olean-12-ene-3 beta,28-diol
Type Analytical Standard
Quantity 10 mg
Formula Weight 442.72
Percent Purity ≥97% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.