Learn More
Apexbio Technology LLC Erythromycin 114-07-8 5g
SDP
Catalog No. 5000092726
Encompass_Preferred
Erythromycin (114-07-8) is a small-molecule inhibitor targeting the bacterial 50S ribosomal subunit It is designed to inhibit bacterial protein translation thereby suppressing bacterial growth Erythromycin exerts its biological activity primarily through reversible binding to the bacterial 50S ribosomal subunit inhibiting protein synthesis Reported IC50 values typically range from 0 01 to 1 0 g/mL depending on bacterial species and strain susceptibility Based on these pharmacological properties erythromycin holds research potential in microbiological assays for ribosomal function translational mechanisms bacterial resistance studies antimicrobial susceptibility testing and cell biology research involving protein translation inhibition
Catalog No. 50-000-92726 Supplier Apexbio Technology LLC Supplier No. B19395000
Erythromycin (114-07-8) is a small-molecule inhibitor targeting the bacterial 50S ribosomal subunit It is designed to inhibit bacterial protein translation thereby suppressing bacterial growth Erythromycin exerts its biological activity primarily through reversible binding to the bacterial 50S ribosomal subunit inhibiting protein synthesis Reported IC50 values typically range from 0 01 to 1 0 g/mL depending on bacterial species and strain susceptibility Based on these pharmacological properties erythromycin holds research potential in microbiological assays for ribosomal function translational mechanisms bacterial resistance studies antimicrobial susceptibility testing and cell biology research involving protein translation inhibition