Learn More
Medchemexpress LLC Erythromycin | 114-07-8 | 98.0% | 1 ML
SDP
Catalog No. 5000405638
Encompass_Preferred
Erythromycin is a macrolide antibiotic derived from Streptomyces erythreus, offering a broad spectrum of antimicrobial activity. It functions by binding to bacterial 50S ribosomal subunits, thereby inhibiting RNA-dependent protein synthesis through the blockage of transpeptidation and/or translocation reactions. Beyond its antibiotic properties, Erythromycin has shown promise with antitumor and neuroprotective effects in various research areas.
  • Binds to bacterial 50S ribosomal subunits
  • Inhibits RNA-dependent protein synthesis
  • Blocks transpeptidation and/or translocation reactions
  • Demonstrates antitumor effects
  • Shows neuroprotective effects
Catalog No. 50-004-05638 Supplier Medchemexpress LLC Supplier No. HYB022010MM/1ML
Erythromycin is a macrolide antibiotic derived from Streptomyces erythreus, offering a broad spectrum of antimicrobial activity. It functions by binding to bacterial 50S ribosomal subunits, thereby inhibiting RNA-dependent protein synthesis through the blockage of transpeptidation and/or translocation reactions. Beyond its antibiotic properties, Erythromycin has shown promise with antitumor and neuroprotective effects in various research areas.
  • Binds to bacterial 50S ribosomal subunits
  • Inhibits RNA-dependent protein synthesis
  • Blocks transpeptidation and/or translocation reactions
  • Demonstrates antitumor effects
  • Shows neuroprotective effects