Learn More
Medchemexpress LLC Erythromycin | 114-07-8 | 98.0% | 10 G
SDP
Catalog No. 5000405492
Encompass_Preferred
Erythromycin is a macrolide antibiotic produced by the actinomycete Streptomyces erythreus with a broad spectrum of antimicrobial activity. It binds to bacterial 50S ribosomal subunits and inhibits RNA-dependent protein synthesis by blocking transpeptidation and/or translocation reactions, without affecting nucleic acid synthesis. Erythromycin also exhibits antitumor and neuroprotective effects in different fields of research.
  • Binds to bacterial 50S ribosomal subunits
  • Inhibits RNA-dependent protein synthesis
  • Blocks transpeptidation and/or translocation reactions
  • Exhibits broad spectrum antimicrobial activity
  • Demonstrates antitumor effects
  • Provides neuroprotective effects
Catalog No. 50-004-05492 Supplier Medchemexpress LLC Supplier No. HYB022010G
Erythromycin is a macrolide antibiotic produced by the actinomycete Streptomyces erythreus with a broad spectrum of antimicrobial activity. It binds to bacterial 50S ribosomal subunits and inhibits RNA-dependent protein synthesis by blocking transpeptidation and/or translocation reactions, without affecting nucleic acid synthesis. Erythromycin also exhibits antitumor and neuroprotective effects in different fields of research.
  • Binds to bacterial 50S ribosomal subunits
  • Inhibits RNA-dependent protein synthesis
  • Blocks transpeptidation and/or translocation reactions
  • Exhibits broad spectrum antimicrobial activity
  • Demonstrates antitumor effects
  • Provides neuroprotective effects