Learn More
Medchemexpress LLC Erythromycin | 114-07-8 | 98.0% | 5 G
SDP
Catalog No. 5000406119
Encompass_Preferred
Erythromycin is a macrolide antibiotic produced by the actinomycete *Streptomyces erythreus*, possessing a broad spectrum of antimicrobial activity. It binds to bacterial 50S ribosomal subunits, inhibiting RNA-dependent protein synthesis. It also demonstrates antitumor and neuroprotective effects.
  • Inhibits RNA-dependent protein synthesis
  • Demonstrates antitumor effects
  • Exhibits neuroprotective effects
  • Macrolide antibiotic
  • Broad spectrum antimicrobial activity
Catalog No. 50-004-06119 Supplier Medchemexpress LLC Supplier No. HYB02205G
Erythromycin is a macrolide antibiotic produced by the actinomycete *Streptomyces erythreus*, possessing a broad spectrum of antimicrobial activity. It binds to bacterial 50S ribosomal subunits, inhibiting RNA-dependent protein synthesis. It also demonstrates antitumor and neuroprotective effects.
  • Inhibits RNA-dependent protein synthesis
  • Demonstrates antitumor effects
  • Exhibits neuroprotective effects
  • Macrolide antibiotic
  • Broad spectrum antimicrobial activity