Learn More
Medchemexpress LLC Erythromycin | 114-07-8 | MFCD00801815 | 98.0% | 733.93 | 500 MG
SDP
Catalog No. 5000365921
Encompass_Preferred
Erythromycin is a macrolide antibiotic produced by the actinomycete *Streptomyces erythreus*, exhibiting a broad spectrum of antimicrobial activity. It functions by binding to bacterial 50S ribosomal subunits, thereby inhibiting RNA-dependent protein synthesis through blockage of transpeptidation and/or translocation reactions, without affecting nucleic acid synthesis. Erythromycin has also demonstrated antitumor and neuroprotective effects in various research fields.
  • Macrolide antibiotic
  • Produced by the actinomycete *Streptomyces erythreus*
  • Exhibits broad spectrum antimicrobial activity
  • Inhibits RNA-dependent protein synthesis by binding to bacterial 50S ribosomal subunits
  • Demonstrates antitumor effects
  • Shows neuroprotective effects
Catalog No. 50-003-65921 Supplier Medchemexpress LLC Supplier No. 500MGHYB0220
Erythromycin is a macrolide antibiotic produced by the actinomycete *Streptomyces erythreus*, exhibiting a broad spectrum of antimicrobial activity. It functions by binding to bacterial 50S ribosomal subunits, thereby inhibiting RNA-dependent protein synthesis through blockage of transpeptidation and/or translocation reactions, without affecting nucleic acid synthesis. Erythromycin has also demonstrated antitumor and neuroprotective effects in various research fields.
  • Macrolide antibiotic
  • Produced by the actinomycete *Streptomyces erythreus*
  • Exhibits broad spectrum antimicrobial activity
  • Inhibits RNA-dependent protein synthesis by binding to bacterial 50S ribosomal subunits
  • Demonstrates antitumor effects
  • Shows neuroprotective effects