Learn More
Medchemexpress LLC Erythromycin-13C2 | 202114-52-1 | 1 MG
SDP
Catalog No. 5000405693
Encompass_Preferred
Erythromycin-13C2 is a macrolide antibiotic used to treat respiratory, skin, and chlamydia infections. It is for research use only and not sold to patients. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-05693 Supplier Medchemexpress LLC Supplier No. HYB0220S31MG
Erythromycin-13C2 is a macrolide antibiotic used to treat respiratory, skin, and chlamydia infections. It is for research use only and not sold to patients. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.