Learn More
Medchemexpress LLC Erythromycin-13C2 | 202114-52-1 | 5 MG
SDP
Catalog No. 5000405777
Encompass_Preferred
Erythromycin-13C2 is a macrolide antibiotic that can be used to treat respiratory infections, skin infections, and chlamydia infections.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-05777 Supplier Medchemexpress LLC Supplier No. HYB0220S35MG
Erythromycin-13C2 is a macrolide antibiotic that can be used to treat respiratory infections, skin infections, and chlamydia infections.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS