Learn More
Medchemexpress LLC Erythromycin, 2'-propanoate, dodecyl sulfate (1:1) | 3521-62-8 | 50 MG
SDP
Catalog No. 5000418643
Encompass_Preferred
Erythromycin estolate is a macrolide antibiotic derivative that is used to treat various bacterial infections. Intended for research use only, it has been noted to cause liver injury, primarily cholestatic hepatitis, and inhibits several flaviviruses such as Zika and dengue viruses, and human coronavirus OC43.
  • Macrolide antibiotic derivative
  • Used to treat various bacterial infections
  • Causes liver injury, primarily cholestatic hepatitis
  • Inhibits several flaviviruses including Zika virus, dengue virus, yellow fever virus, and human coronavirus OC43
  • Inactivates viral particles by compromising the integrity of the viral membrane
  • Reduces virus titer of HCoV-OC43 with an IC50 of 1.40 μM
Catalog No. 50-004-18643 Supplier Medchemexpress LLC Supplier No. HYN712150MG
Erythromycin estolate is a macrolide antibiotic derivative that is used to treat various bacterial infections. Intended for research use only, it has been noted to cause liver injury, primarily cholestatic hepatitis, and inhibits several flaviviruses such as Zika and dengue viruses, and human coronavirus OC43.
  • Macrolide antibiotic derivative
  • Used to treat various bacterial infections
  • Causes liver injury, primarily cholestatic hepatitis
  • Inhibits several flaviviruses including Zika virus, dengue virus, yellow fever virus, and human coronavirus OC43
  • Inactivates viral particles by compromising the integrity of the viral membrane
  • Reduces virus titer of HCoV-OC43 with an IC50 of 1.40 μM