Learn More
Cayman Chemical Erythromycin 2'-Propionate, Purity: ≥95%, CAS: 134-36-1, 25 mg
SDP
Catalog No. 502990880
Encompass_Preferred
An antibiotic; active against S. aureus at 1 µg/ml; inhibits protein synthesis in a cell-free assay at 1 mM
Catalog No. 50-299-0880 Supplier Cayman Chemical Supplier No. 3173925 MG
An antibiotic; active against S. aureus at 1 µg/ml; inhibits protein synthesis in a cell-free assay at 1 mM