Learn More
Cayman Chemical Erythromycin 2'-Propionate, Purity: ≥95%, CAS: 134-36-1, 5 mg
SDP
Catalog No. 502992200
Encompass_Preferred
An antibiotic; active against S. aureus at 1 µg/ml; inhibits protein synthesis in a cell-free assay at 1 mM
Catalog No. 50-299-2200 Supplier Cayman Chemical Supplier No. 317395 MG
An antibiotic; active against S. aureus at 1 µg/ml; inhibits protein synthesis in a cell-free assay at 1 mM