Learn More
Alkali Scientific Erythromycin, 25 G
SDP
Catalog No. 5000660315
Encompass_Preferred
Erythromycin is a macrolide antibiotic widely used in laboratory applications for its ability to inhibit bacterial protein synthesis. It is effective against a range of Gram-positive organisms and selected Gram-negative bacteria, making it suitable for antimicrobial testing and selective culture applications. Supplied as a high-purity powder, it is commonly incorporated into microbiological media to control bacterial growth or study resistance mechanisms. The 25 g size provides a convenient bulk option for laboratories performing frequent testing or large-scale experimental work. Its consistent quality ensures reliable performance in microbiology, pharmaceutical development, and biochemical research settings.
Catalog No. 50-006-60315 Supplier Alkali Scientific Supplier No. ERY002CP
Erythromycin is a macrolide antibiotic widely used in laboratory applications for its ability to inhibit bacterial protein synthesis. It is effective against a range of Gram-positive organisms and selected Gram-negative bacteria, making it suitable for antimicrobial testing and selective culture applications. Supplied as a high-purity powder, it is commonly incorporated into microbiological media to control bacterial growth or study resistance mechanisms. The 25 g size provides a convenient bulk option for laboratories performing frequent testing or large-scale experimental work. Its consistent quality ensures reliable performance in microbiology, pharmaceutical development, and biochemical research settings.