Learn More
Bioworld Erythromycin, 10 g
Supplier:  Bioworld 405100001
Encompass_Preferred
Erythromycin A macrolide antibiotic against Gram-positive bacteria. It binds to 50S ribosomal subunit and blocks bacterial protein synthesis.USP GradeA macrolide antibiotic protein synthesis inhibitorPotency >850 μg/mgWater <10.0%Residue on ignition <2.0%.For in vitro lab use only. Not for human use or consumption.Appearance: SolidBoiling Point : 818.4 °C at 760 mmHg (Predicted)Density : 1.2 g/cm3 (Predicted)Melting Point: 135-140 °C (lit.)RTECS Number: KF4375000
Catalog No. 50-255-953