Learn More
Alkali Scientific Erythromycin, 50 Grams
SDP
Catalog No. 5000660190
Encompass_Preferred
Erythromycin (50 g) is a broad-spectrum antibiotic that is particularly effective against Gram-positive bacteria and is commonly used to treat respiratory, skin, and soft tissue infections. As a macrolide antibiotic, erythromycin works by inhibiting bacterial protein synthesis through binding to the 50S ribosomal subunit. This prevents bacterial growth and replication, effectively treating infections caused by susceptible organisms. Erythromycin is used in both clinical and research settings, often prescribed as an alternative for patients allergic to penicillin. This 50 g quantity is ideal for laboratories conducting antimicrobial susceptibility testing and for clinical formulations requiring larger doses of the antibiotic.
Catalog No. 50-006-60190 Supplier Alkali Scientific Supplier No. ERY003CP
Erythromycin (50 g) is a broad-spectrum antibiotic that is particularly effective against Gram-positive bacteria and is commonly used to treat respiratory, skin, and soft tissue infections. As a macrolide antibiotic, erythromycin works by inhibiting bacterial protein synthesis through binding to the 50S ribosomal subunit. This prevents bacterial growth and replication, effectively treating infections caused by susceptible organisms. Erythromycin is used in both clinical and research settings, often prescribed as an alternative for patients allergic to penicillin. This 50 g quantity is ideal for laboratories conducting antimicrobial susceptibility testing and for clinical formulations requiring larger doses of the antibiotic.