Learn More
Selleck Chemical LLC Erythromycin-50mg
SDP
Catalog No. 502389643
Encompass_Preferred
Erythromycin (E-Mycin) is a macrolide antibiotic that has an antimicrobial spectrum similar to or slightly wider than that of penicillin (IC50=1.5 μg/ml).
Catalog No. 50-238-9643 Supplier Selleck Chemical LLC Supplier No. S163550MG
Erythromycin (E-Mycin) is a macrolide antibiotic that has an antimicrobial spectrum similar to or slightly wider than that of penicillin (IC50=1.5 μg/ml).