Learn More
Cayman Chemical Erythromycin, Purity: ≥98%, CAS: 114-07-8, 5 g
SDP
Catalog No. 502969284
Encompass_Preferred
A macrolide antibiotic; inhibits 50S ribosome activity; active against a host of bacterial genera, including Streptococcus, Staphylococcus, and Haemophilus (MIC90s = 0.015-2.0 mg/l); dose-dependently inhibits the growth of S. aureus in a mouse model of thigh infection at 10-40 mg/kg; inhibits CYP3A4 (IC50s = 33 and 27.3 µM for α-hydroxytriazolam and 4-hydroxytriazolam formation, respectively)
Catalog No. 50-296-9284 Supplier Cayman Chemical Supplier No. 164865 G
A macrolide antibiotic; inhibits 50S ribosome activity; active against a host of bacterial genera, including Streptococcus, Staphylococcus, and Haemophilus (MIC90s = 0.015-2.0 mg/l); dose-dependently inhibits the growth of S. aureus in a mouse model of thigh infection at 10-40 mg/kg; inhibits CYP3A4 (IC50s = 33 and 27.3 µM for α-hydroxytriazolam and 4-hydroxytriazolam formation, respectively)