Learn More
Enzo Life Sciences Erythromycin (5g). CAS: 114-07-8
SDP
Catalog No. 502009007
Encompass_Preferred
Macrolide antibiotic. Inhibits protein synthesis (elongation) at the transpeptidation step. Binds to the 23S RNA of the 50S ribosomal subunit. Effective against Gram-negative and Gram-positive bacteria. Purity: ≥85% (Assay on dry basis). Appearance: White to off-white powder. Solubility: Soluble in water (50mg/ml), 100% ethanol or methanol. Long Term Storage: -20°C.
Catalog No. 50-200-9007 Supplier Enzo Life Sciences Supplier No. ALX380274G005
Macrolide antibiotic. Inhibits protein synthesis (elongation) at the transpeptidation step. Binds to the 23S RNA of the 50S ribosomal subunit. Effective against Gram-negative and Gram-positive bacteria. Purity: ≥85% (Assay on dry basis). Appearance: White to off-white powder. Solubility: Soluble in water (50mg/ml), 100% ethanol or methanol. Long Term Storage: -20°C.