Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erythromycin, ∽98%, MP Biomedicals™

Catalog No. MP219019705 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219019705 5 g
ICN19019701 1 g
MP219019725 25 g
ICN19019780 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MP219019705 Supplier MP Biomedicals, Inc Supplier No. 0219019705
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity antibiotic with>98% purity for research and laboratory applications.

  • It Used in cell culture applications at approximately 100 mg/liter.
  • Depending upon the strain of bacteria, erythromycin has been used between 50 and 200 mg/L to control bacterial growth.
  • Erythromycin resistance can be induced in bacteria.
  • Erythromycin is a macrolide antibiotic that bind to the 50S ribosomal subunit and inhibit protein synthesis.
  • This is an antimicrobial substance produced by the growth of certain strains of Streptomyces erythreus.
  • It is a mixture consisting largely of erythromycin A with lesser amounts of erythromycins B and C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114-07-8
Molecular Formula C37H67NO13
Molecular Weight (g/mol) 733.94
MDL Number MFCD00084654
InChI Key ULGZDMOVFRHVEP-RWJQBGPGSA-N
IUPAC Name (3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-6-{[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy}-14-ethyl-7,12,13-trihydroxy-4-{[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy}-3,5,7,9,11,13-hexamethyl-1-oxacyclotetradecane-2,10-dione
SMILES CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]2O[C@H](C)C[C@@H]([C@H]2O)N(C)C)[C@](C)(O)C[C@@H](C)C(=O)[C@H](C)[C@@H](O)[C@]1(C)O

Specifications

Type Erythromycin
pH 8.0 to 10.5 (70% aq soln)
Melting Point 135 to 140°C (literature)
Extinction Coefficient EM = 50 (280 nm, pH 6.3)
Potency ≥850 µg/mg
Quantity 5 g
Optical Rotation -71° to -78° (c=2, ethanol)
Beilstein 75279
Synonym E- mycin, Erycin, Robimysin, Erythrocin
Solubility Information Slightly soluble in water (∽2mg/mL); Moderately soluble in ether, ethylene dichloride, amyl acetate; freely soluble in alcohols (ethanol - 50mg/mL), acetone, chloroform, acetonitrile, ethyl acetate. Soluble in 2 M Hydrochloric acid (50mg/mL).
Percent Purity 98%
Grade ∽98%
Show More Show Less
RTECSNumber KF4375000
storageNote1 Store at Room Temperature (15-30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.