Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Erythromycin A 9-methoxime | 53066-26-5 | MFCD00864895 | 97.6% | 763.0 g/mol | C38H70N2O13 | 25 MG
SDP

Catalog No. 5000243799
Encompass_Preferred

Lexithromycin (erythromycin A 9-methoxime) is a macrolide antibiotic derivative supplied for laboratory research and analytical use. It demonstrates antibacterial activity and is provided as a solid material suitable for method development and biological assays.

  • Used as a research reagent in antibacterial studies.
  • Available as an analytical standard for method development and quality control.
  • CAS number 53066-26-5.
  • Purity approximately 97.6% as supplied.
  • Solid, white to off-white appearance.
  • Molecular formula C38H70N2O13; molar mass 763.0 g/mol.
  • Offered in small laboratory package sizes (for example, 25 mg) suitable for analytical work.

Catalog No. 50-002-43799 Supplier Medchemexpress LLC Supplier No. HY10593225MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lexithromycin (erythromycin A 9-methoxime) is a macrolide antibiotic derivative supplied for laboratory research and analytical use. It demonstrates antibacterial activity and is provided as a solid material suitable for method development and biological assays.

  • Used as a research reagent in antibacterial studies.
  • Available as an analytical standard for method development and quality control.
  • CAS number 53066-26-5.
  • Purity approximately 97.6% as supplied.
  • Solid, white to off-white appearance.
  • Molecular formula C38H70N2O13; molar mass 763.0 g/mol.
  • Offered in small laboratory package sizes (for example, 25 mg) suitable for analytical work.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.