Learn More
TARGETMOL CHEMICALS INC ERYTHROMYCIN A ENOL ETHE 5MG
SDP
Catalog No. 502581024
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Erythromycin A enol ether is a degradation product of Erythromycin A which is a macrolide antibiotic. purity: 99%
Catalog No. 50-258-1024 Supplier TARGETMOL CHEMICALS INC Supplier No. T21572
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Erythromycin A enol ether is a degradation product of Erythromycin A which is a macrolide antibiotic. purity: 99%