Learn More
Cayman Chemical Erythromycin A enol ether, Purity: ≥95%, CAS: 33396-29-1, 1 mg
SDP
Catalog No. 502966959
Encompass_Preferred
A decomposition product of the macrolide antibiotic, erythromycin A that has been identified as a β-turn mimic of the peptide hormone motilin; used to determine the binding characteristics of ligands of the motilin receptor
Catalog No. 50-296-6959 Supplier Cayman Chemical Supplier No. 171911 MG
A decomposition product of the macrolide antibiotic, erythromycin A that has been identified as a β-turn mimic of the peptide hormone motilin; used to determine the binding characteristics of ligands of the motilin receptor