Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Erythromycin A enol ether | 33396-29-1 | 98.9% | 1 MG
SDP

Catalog No. 5000231165
Encompass_Preferred

Erythromycin A enol ether is an acidic degradation product of erythromycin A supplied as a high-purity analytical standard for research and impurity profiling. It is provided as a solid in small package sizes suitable for analytical workflows.

  • Analytical standard for impurity identification and method development.
  • Acidic degradation product of erythromycin A with no antibacterial activity.
  • High reported purity (98.86%).
  • Molecular formula C37H65NO12, molecular weight 715.91 g/mol.
  • Available in small quantities down to 1 mg for analytical use.
  • Physical form: solid; intended for research use only.

Catalog No. 50-002-31165 Supplier Medchemexpress LLC Supplier No. HY1222071MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Erythromycin A enol ether is an acidic degradation product of erythromycin A supplied as a high-purity analytical standard for research and impurity profiling. It is provided as a solid in small package sizes suitable for analytical workflows.

  • Analytical standard for impurity identification and method development.
  • Acidic degradation product of erythromycin A with no antibacterial activity.
  • High reported purity (98.86%).
  • Molecular formula C37H65NO12, molecular weight 715.91 g/mol.
  • Available in small quantities down to 1 mg for analytical use.
  • Physical form: solid; intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.