Learn More
Medchemexpress LLC Erythromycin A enol ether | 33396-29-1 | 98.9% | 100 MG
SDP
Catalog No. 5000230864
Encompass_Preferred
Erythromycin A enol ether is an analytical-grade degradation product of the macrolide antibiotic erythromycin A supplied as a solid standard for laboratory research. It is used in impurity profiling, degradation studies, and analytical method development, and it does not exhibit antibacterial activity.
  • Analytical standard of erythromycin A degradation product.
  • Purity 98.9% suitable for chromatographic and spectrometric analysis.
  • Used for impurity profiling, degradation studies, and method development.
  • Supplied as a solid standard in multiple sizes for flexible experimental use.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-30864 Supplier Medchemexpress LLC Supplier No. HY122207100MG
Erythromycin A enol ether is an analytical-grade degradation product of the macrolide antibiotic erythromycin A supplied as a solid standard for laboratory research. It is used in impurity profiling, degradation studies, and analytical method development, and it does not exhibit antibacterial activity.
  • Analytical standard of erythromycin A degradation product.
  • Purity 98.9% suitable for chromatographic and spectrometric analysis.
  • Used for impurity profiling, degradation studies, and method development.
  • Supplied as a solid standard in multiple sizes for flexible experimental use.
  • For research use only; not for human or veterinary use.