Learn More
Medchemexpress LLC Erythromycin A enol ether | 33396-29-1 | 98.9% | 25 MG
SDP
Catalog No. 5000232133
Encompass_Preferred
Erythromycin A enol ether is an acidic degradation product of the macrolide antibiotic erythromycin A. Supplied as an analytical standard for research purposes, it has no antibacterial activity and is provided as a white to off-white solid with high purity suitable for analytical and impurity profiling.
  • Analytical standard for reference and impurity profiling.
  • High purity (98.9%) for reliable chromatographic analysis.
  • Available in small milligram pack sizes for research workflows.
  • Stable when stored at -20°C under nitrogen for long-term storage.
  • For research use only; not for human or clinical applications.
Catalog No. 50-002-32133 Supplier Medchemexpress LLC Supplier No. HY12220725MG
Erythromycin A enol ether is an acidic degradation product of the macrolide antibiotic erythromycin A. Supplied as an analytical standard for research purposes, it has no antibacterial activity and is provided as a white to off-white solid with high purity suitable for analytical and impurity profiling.
  • Analytical standard for reference and impurity profiling.
  • High purity (98.9%) for reliable chromatographic analysis.
  • Available in small milligram pack sizes for research workflows.
  • Stable when stored at -20°C under nitrogen for long-term storage.
  • For research use only; not for human or clinical applications.