Learn More
Medchemexpress LLC Erythromycin A enol ether | 33396-29-1 | 98.9% | 5 MG
SDP
Catalog No. 5000233766
Encompass_Preferred
Erythromycin A enol ether is an acidic degradation product of erythromycin A that lacks antibacterial activity and is provided for research use only.
  • Acidic degradation product of erythromycin A with no antibacterial effect.
  • High reported purity of 98.9% and molecular weight of 715.91 g/mol.
  • Available in small milligram quantities suitable for analytical and stability studies.
  • Analytical documentation available: data sheet, COA, H-NMR, LC-MS, and SDS.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-33766 Supplier Medchemexpress LLC Supplier No. HY1222075MG
Erythromycin A enol ether is an acidic degradation product of erythromycin A that lacks antibacterial activity and is provided for research use only.
  • Acidic degradation product of erythromycin A with no antibacterial effect.
  • High reported purity of 98.9% and molecular weight of 715.91 g/mol.
  • Available in small milligram quantities suitable for analytical and stability studies.
  • Analytical documentation available: data sheet, COA, H-NMR, LC-MS, and SDS.
  • For research use only; not for human or clinical use.