Learn More
Medchemexpress LLC Erythromycin A enol ether | 33396-29-1 | 98.9% | 50 MG
SDP
Catalog No. 5000232134
Encompass_Preferred
Erythromycin A enol ether is an acidic degradation product of the macrolide antibiotic erythromycin A. It lacks antibacterial activity and is supplied as an analytical reference standard for impurity identification, stability studies, and structure-activity investigations.
  • Analytical standard for HPLC and LC-MS methods.
  • Reference material for impurity profiling and degradation studies.
  • High listed purity suitable for analytical use.
  • Available in multiple small pack sizes for laboratory testing.
Catalog No. 50-002-32134 Supplier Medchemexpress LLC Supplier No. HY12220750MG
Erythromycin A enol ether is an acidic degradation product of the macrolide antibiotic erythromycin A. It lacks antibacterial activity and is supplied as an analytical reference standard for impurity identification, stability studies, and structure-activity investigations.
  • Analytical standard for HPLC and LC-MS methods.
  • Reference material for impurity profiling and degradation studies.
  • High listed purity suitable for analytical use.
  • Available in multiple small pack sizes for laboratory testing.