Learn More
Cayman Chemical Erythromycin A N-oxide, Purity: ≥95%, CAS: 992-65-4, 25 mg
SDP
Catalog No. 502994665
Encompass_Preferred
A potential impurity in commercial preparations of erythromycin; a precursor in the synthesis of clarithromycin
Catalog No. 50-299-4665 Supplier Cayman Chemical Supplier No. 2364225 MG
A potential impurity in commercial preparations of erythromycin; a precursor in the synthesis of clarithromycin