Learn More
Cayman Chemical ErythromycIn C 500ug
SDP
Catalog No. 502994241
Encompass_Preferred
An intermediate in the biosynthesis of erythromycin; a potential impurity in commercial preparations of erythromycin; half as active or less than erythromycin A or B against most Gram-positive and Gram-negative bacteria tested
Catalog No. 50-299-4241 Supplier Cayman Chemical Supplier No. 23478500 UG
An intermediate in the biosynthesis of erythromycin; a potential impurity in commercial preparations of erythromycin; half as active or less than erythromycin A or B against most Gram-positive and Gram-negative bacteria tested