Learn More
Cayman Chemical Erythromycin C, Purity: ≥85%, CAS: 1675-02-1, 5 mg
SDP
Catalog No. 502994598
Encompass_Preferred
An intermediate in the biosynthesis of erythromycin; a potential impurity in commercial preparations of erythromycin; half as active or less than erythromycin A or B against most Gram-positive and Gram-negative bacteria tested
Catalog No. 50-299-4598 Supplier Cayman Chemical Supplier No. 234785 MG
An intermediate in the biosynthesis of erythromycin; a potential impurity in commercial preparations of erythromycin; half as active or less than erythromycin A or B against most Gram-positive and Gram-negative bacteria tested