Learn More
Medchemexpress LLC Erythromycin cyclocarbonate | 55224-05-0 | 759.92 | C38H65NO14 | 1 MG
SDP
Catalog No. 5000213745
Encompass_Preferred
Davercin (erythromycin cyclocarbonate) is a semi-synthetic macrolide antibiotic derivative used in research to study antibacterial activity and mechanisms of protein synthesis inhibition. It is active primarily against Gram-positive organisms and some Gram-negative strains, and is provided as a characterized reagent for microbiology and pharmacology assays.
  • Derivative of erythromycin with macrolide activity.
  • Active against Gram-positive and some Gram-negative microorganisms.
  • Useful for research on bacterial protein synthesis inhibition.
  • Characterized chemically: formula C38H65NO14, molecular weight 759.92 g/mol.
  • Supplied in small research quantities suitable for assay development and characterization.
Catalog No. 50-002-13745 Supplier Medchemexpress LLC Supplier No. HY1005841MG
Davercin (erythromycin cyclocarbonate) is a semi-synthetic macrolide antibiotic derivative used in research to study antibacterial activity and mechanisms of protein synthesis inhibition. It is active primarily against Gram-positive organisms and some Gram-negative strains, and is provided as a characterized reagent for microbiology and pharmacology assays.
  • Derivative of erythromycin with macrolide activity.
  • Active against Gram-positive and some Gram-negative microorganisms.
  • Useful for research on bacterial protein synthesis inhibition.
  • Characterized chemically: formula C38H65NO14, molecular weight 759.92 g/mol.
  • Supplied in small research quantities suitable for assay development and characterization.