Learn More
Medchemexpress LLC Erythromycin cyclocarbonate | 55224-05-0 | 98.0% | 759.92 g/mol | C38H65NO14 | 25 MG
SDP
Catalog No. 5000213662
Encompass_Preferred
Davercin (erythromycin cyclocarbonate) is a semi-synthetic derivative of erythromycin used as an antibacterial research reagent. It inhibits bacterial protein synthesis and is active against Gram-positive and some Gram-negative organisms. Supplied as a solid for laboratory studies and analytical reference.
  • Derivative of erythromycin with antibacterial activity.
  • Active against Gram-positive and some Gram-negative organisms.
  • High purity (98.0%).
  • Molecular weight 759.92 g/mol and formula C38H65NO14.
  • White to off-white solid suitable for laboratory use.
  • Available in small research quantities such as 25 MG.
Catalog No. 50-002-13662 Supplier Medchemexpress LLC Supplier No. HY10058425MG
Davercin (erythromycin cyclocarbonate) is a semi-synthetic derivative of erythromycin used as an antibacterial research reagent. It inhibits bacterial protein synthesis and is active against Gram-positive and some Gram-negative organisms. Supplied as a solid for laboratory studies and analytical reference.
  • Derivative of erythromycin with antibacterial activity.
  • Active against Gram-positive and some Gram-negative organisms.
  • High purity (98.0%).
  • Molecular weight 759.92 g/mol and formula C38H65NO14.
  • White to off-white solid suitable for laboratory use.
  • Available in small research quantities such as 25 MG.