Learn More
Medchemexpress LLC Erythromycin cyclocarbonate | 55224-05-0 | MFCD01729176 | >97.0% | 759.92 | C38H65NO14 | 50 MG
SDP
Catalog No. 5000213925
Encompass_Preferred
Davercin (erythromycin cyclocarbonate) is a macrolide antibiotic derivative of erythromycin used as a research compound. It inhibits bacterial protein synthesis and is active against Gram-positive and some Gram-negative microorganisms. Supplied as a powder, it is intended for in vitro antibacterial studies and medicinal chemistry research.
  • Macrolide antibiotic derivative suitable for research use.
  • Inhibits bacterial protein synthesis by targeting the 50S ribosomal subunit.
  • Active against Gram-positive and some Gram-negative microorganisms.
  • Supplied as a powder for flexible formulation and dosing.
  • Defined storage conditions to maintain stability.
  • Available in small quantities tailored for laboratory studies.
Catalog No. 50-002-13925 Supplier Medchemexpress LLC Supplier No. HY10058450MG
Davercin (erythromycin cyclocarbonate) is a macrolide antibiotic derivative of erythromycin used as a research compound. It inhibits bacterial protein synthesis and is active against Gram-positive and some Gram-negative microorganisms. Supplied as a powder, it is intended for in vitro antibacterial studies and medicinal chemistry research.
  • Macrolide antibiotic derivative suitable for research use.
  • Inhibits bacterial protein synthesis by targeting the 50S ribosomal subunit.
  • Active against Gram-positive and some Gram-negative microorganisms.
  • Supplied as a powder for flexible formulation and dosing.
  • Defined storage conditions to maintain stability.
  • Available in small quantities tailored for laboratory studies.