Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Erythromycin cyclocarbonate | 55224-05-0 | MFCD01729176 | ≥98.0% | 759.92 | C38H65NO14 | 100 MG
SDP

Catalog No. 5000214711
Encompass_Preferred

Davercin (erythromycin cyclocarbonate) is a macrolide antibiotic derivative used as a research reagent. It inhibits bacterial protein synthesis and is reported to be active against Gram-positive and some Gram-negative microorganisms. The product is supplied in solid and solution formats with manufacturer guidance for solvent preparation and handling.

  • Derivative of erythromycin with antibacterial activity against Gram-positive and some Gram-negative organisms.
  • Molecular formula C38H65NO14 and molecular weight about 759.92 g/mol.
  • CAS number 55224-05-0 for unambiguous chemical identification.
  • Available in solid sizes from 1 mg to 500 mg and as 10 mM solutions in DMSO.
  • Soluble in DMSO for preparing stock solutions; limited solubility in water.
  • Intended for laboratory research use; follow the safety data sheet and handling instructions.

Catalog No. 50-002-14711 Supplier Medchemexpress LLC Supplier No. HY100584100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Davercin (erythromycin cyclocarbonate) is a macrolide antibiotic derivative used as a research reagent. It inhibits bacterial protein synthesis and is reported to be active against Gram-positive and some Gram-negative microorganisms. The product is supplied in solid and solution formats with manufacturer guidance for solvent preparation and handling.

  • Derivative of erythromycin with antibacterial activity against Gram-positive and some Gram-negative organisms.
  • Molecular formula C38H65NO14 and molecular weight about 759.92 g/mol.
  • CAS number 55224-05-0 for unambiguous chemical identification.
  • Available in solid sizes from 1 mg to 500 mg and as 10 mM solutions in DMSO.
  • Soluble in DMSO for preparing stock solutions; limited solubility in water.
  • Intended for laboratory research use; follow the safety data sheet and handling instructions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.