Learn More
Medchemexpress LLC Erythromycin cyclocarbonate | 55224-05-0 | MFCD01729176 | >98.0% | 759.92 | C38H65NO14 | 5 MG
SDP
Catalog No. 5000213924
Encompass_Preferred
Davercin (erythromycin cyclocarbonate) is a semi-synthetic macrolide derivative of erythromycin used as a research reagent. It is reported to inhibit bacterial protein synthesis by binding the 50S ribosomal subunit and shows activity against Gram-positive and some Gram-negative organisms, making it useful for microbiology and antibiotic studies.
  • Macrolide antibiotic derivative active against Gram-positive and some Gram-negative bacteria.
  • Inhibits protein synthesis through 50S ribosomal binding.
  • Suitable as a reference standard or reagent in microbiology and antibiotic research.
  • Provided in small quantities for preparing concentrated stock solutions (for example, 1 mg, 5 mg).
  • Characterized by CAS 55224-05-0 and molecular formula C38H65NO14.
Catalog No. 50-002-13924 Supplier Medchemexpress LLC Supplier No. HY1005845MG
Davercin (erythromycin cyclocarbonate) is a semi-synthetic macrolide derivative of erythromycin used as a research reagent. It is reported to inhibit bacterial protein synthesis by binding the 50S ribosomal subunit and shows activity against Gram-positive and some Gram-negative organisms, making it useful for microbiology and antibiotic studies.
  • Macrolide antibiotic derivative active against Gram-positive and some Gram-negative bacteria.
  • Inhibits protein synthesis through 50S ribosomal binding.
  • Suitable as a reference standard or reagent in microbiology and antibiotic research.
  • Provided in small quantities for preparing concentrated stock solutions (for example, 1 mg, 5 mg).
  • Characterized by CAS 55224-05-0 and molecular formula C38H65NO14.